Learn More
Selleck Chemical LLC Upadacitinib
SDP
Catalog No. 5000000684
Encompass_Preferred
Upadacitinib is a selective JAK1 inhibitor which demonstrates activity against JAK1 (0 045 M) and JAK2 (0 109 M) with 40 fold selectivity over JAK3 (2 1 M) and 100 fold selectivity over TYK2 (4 7 M) as compared to JAK1
Catalog No. 50-000-00684 Supplier Selleck Chemical LLC Supplier No. S8162100MG
Upadacitinib is a selective JAK1 inhibitor which demonstrates activity against JAK1 (0 045 M) and JAK2 (0 109 M) with 40 fold selectivity over JAK3 (2 1 M) and 100 fold selectivity over TYK2 (4 7 M) as compared to JAK1