Learn More
Medchemexpress LLC HY-19569 50mg , Upadacitinib CAS:1310726-60-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1956950MG
Encompass_Preferred
HY-19569 50mg Upadacitinib CAS: 1310726-60-3 Upadacitinib (ABT-494) is a potent, orally active and selective Janus kinase 1 (JAK1) inhibitor (IC50=43 nM). Upadacitinib (ABT-494) displays approximately 74 fold selective for JAK1 over JAK2 (200 nM) in cellular assays dependent on specific, relevant cytokines. Upadacitinib (ABT-494) can be used for several autoimmune disorders research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1043