Learn More
Selleck Chemical LLC Upadacitinib (ABT-494) 25mg 1310726-60-3
SDP
Supplier:  Selleck Chemical LLC S816225MG
Encompass_Preferred
Upadacitinib (ABT-494) is a selective JAK1 inhibitor which demonstrates activity against JAK1 (0.045 ?M) and JAK2 (0.109 ?M), with > 40 fold selectivity over JAK3 (2.1 ?M) and 100 fold selectivity over TYK2 (4.7 ?M) as compared to JAK1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1828339