Learn More
ACROBiosystems Recombinant Protein;Human uPAR / PLAUR Protein;HEK293;200UG;UPR,uPAR,PLAUR,CD87,MO3,U-PAR
Catalog No. 501658493
Encompass_Preferred
Recombinant Protein;Human uPAR / PLAUR Protein;200UG;Human uPAR, His Tag (UPR-H5226) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # AAH02788).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 32.1 kDa. The protein migrates as 44-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8493 Supplier ACROBiosystems Supplier No. UPRH5226200UG
Recombinant Protein;Human uPAR / PLAUR Protein;200UG;Human uPAR, His Tag (UPR-H5226) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # AAH02788).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 32.1 kDa. The protein migrates as 44-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.