Learn More
Aviva Systems Biology Corp UPB1 Antibody - middle region (ARP42467_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42467_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against UPB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UPB1. Alternate names: BUP1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58235