Learn More
Waters Corp Waters Corporation Standard, Caffeine, 1.0 mg/mL, 2 mL
Supplier: Waters Corp 186006614
This individual caffeine standard is formulated for use with SFC systems, providing a 1.0 mg/mL caffeine concentration in 2-propanol within a 2 mL volume.
- Detection method: UV
- Liquid formulation
- Storage condition: Room temperature
- System type: SFC/UPC2
- Instrument compatibility: SFx
- Quantitative analysis enabled
- Shelf life: 36 months
- Shipping condition: Ambient
- Volume: 2 mL
- Application: System performance
- Units per package: 1
- Contains hazardous material requiring special freight handling; additional charges may apply.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.