Learn More
Waters Corp Waters Gradient Standard, SFC, Mixture, 1 mL
Supplier:  Waters Corp 186006551
Encompass_Preferred
The UPC2 Gradient Standard is a mixture formulated for Supercritical Fluid Chromatography (SFC) use. It contains 1 mg/mL Coumarin, 1 mg/mL Flavone, 2 mg/mL Caffeine, 1 mg/mL Thymine, and 2 mg/mL Prednisone in 2-propanol.
  • Formulated for SFC/UPC2 system type
  • Contains multiple components for gradient analysis
  • Supplied as a liquid formulation
  • Volume of 1 mL
  • Designed for UV detection
  • Suitable for quantitative analysis
  • Has a shelf life of 36 months
  • Stable at room temperature storage
  • Shipped under ambient conditions
  • Ideal for system performance applications
  • Categorized as a reference standard
  • Packaged as one unit
  • Contains hazardous material which requires special freight handling and additional charges
Catalog No. 50-466-311