Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp UPC2 QC Reference Material, 1 mL, Liquid, UV Detection

Catalog No. 502391773 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The UPC2 QC Reference Material provides convergence chromatographic performance data for both chiral and achiral modes. Designed for use with Trefoil and Torus Columns, this four-compound mixture ensures well-separated compounds across a broad elution range and UV detection compatibility.

  • Supports chiral and achiral chromatographic analysis.
  • Designed for use with Trefoil and Torus Columns.
  • Four-compound mixture.
  • Composition: 0.50 mg/mL (±) trans-Stilbene oxide, 0.50 mg/mL Thymine, 0.50 mg/mL Sulfamethoxazole, 0.50 mg/mL Sulfamethizole.
  • 1 mL solution in 75:25 ACN:MeOH.
  • UV detection method.
  • Liquid formulation.
  • Storage condition: 4°C (Fridge).
  • System type: UPC2/SFC.
  • Quantitative.
  • Shelf life: 12 months.
  • Ambient shipping.
  • Volume: 1 mL.
  • Units per package: 1 pk.
  • Hazardous material.

Catalog No. 50-239-1773 Supplier Waters Corp Supplier No. 186007950
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The UPC2 QC Reference Material provides convergence chromatographic performance data for both chiral and achiral modes. Designed for use with Trefoil and Torus Columns, this four-compound mixture ensures well-separated compounds across a broad elution range and UV detection compatibility.

  • Supports chiral and achiral chromatographic analysis.
  • Designed for use with Trefoil and Torus Columns.
  • Four-compound mixture.
  • Composition: 0.50 mg/mL (±) trans-Stilbene oxide, 0.50 mg/mL Thymine, 0.50 mg/mL Sulfamethoxazole, 0.50 mg/mL Sulfamethizole.
  • 1 mL solution in 75:25 ACN:MeOH.
  • UV detection method.
  • Liquid formulation.
  • Storage condition: 4°C (Fridge).
  • System type: UPC2/SFC.
  • Quantitative.
  • Shelf life: 12 months.
  • Ambient shipping.
  • Volume: 1 mL.
  • Units per package: 1 pk.
  • Hazardous material.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.