Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Flavone Standard, 1.0 mg/mL, 2 mL

Catalog No. 50466300 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This individual flavone standard is formulated for use with SFC. It contains 1.0 mg/mL flavone in 2-propanol and comes in a 2 mL volume, making it suitable for system performance applications.

  • Detection method: UV
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: SFC/UPC2
  • Instrument: SFx
  • Quantitative: Yes
  • Shelf life: 36 months
  • Shipping condition: Ambient
  • Volume: 2 mL
  • Type: Reference
  • UNSPSC: 41116107
  • Application: System performance
  • Units per package: 1 pk
  • Contains hazardous material which requires special freight handling.

Catalog No. 50-466-300 Supplier Waters Corp Supplier No. 186006523
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This individual flavone standard is formulated for use with SFC. It contains 1.0 mg/mL flavone in 2-propanol and comes in a 2 mL volume, making it suitable for system performance applications.

  • Detection method: UV
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: SFC/UPC2
  • Instrument: SFx
  • Quantitative: Yes
  • Shelf life: 36 months
  • Shipping condition: Ambient
  • Volume: 2 mL
  • Type: Reference
  • UNSPSC: 41116107
  • Application: System performance
  • Units per package: 1 pk
  • Contains hazardous material which requires special freight handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.