Learn More
Waters Corp UPC2 Standard Flurbiprofen 186006524
Supplier:  Waters Corp 186006524
Encompass_Preferred
Individual flurbiprofen standard formulated for use with SFC. 1.0 mg/mL flurbiprofen in 2-propanol, 2mL
Catalog No. 50-466-301