Learn More
Waters Corp UPC2 Standard Ibuprofen 186006521
Supplier:  Waters Corp 186006521
Encompass_Preferred
Individual ibuprofen standard formulated for use with SFC. 1.0 mg/mL ibuprofen in 2-propanol, 2mL
Catalog No. 50-466-298