Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation UPC2 Standard Mix, UV Detection, 1 mL, 36 Months

Catalog No. 50466295 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The UPC2 Standard Mix is a reference standard comprised of 2 mg/ml each of 3-benzoylpyridine, Cortisone, 4-nitroaniline, and 4,4'-biphenol in methanol solution, designed for system performance applications.

  • Formulated as a liquid for UV detection.
  • Designed for SFC/UPC2 system types.
  • Quantitative reference standard.
  • Volume of 1 mL per package.
  • Shelf life of 36 months when stored at 4°C.
  • Shipped under ambient conditions.
  • Contains hazardous material requiring special freight handling.

Catalog No. 50-466-295 Supplier Waters Corp Supplier No. 186006372
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The UPC2 Standard Mix is a reference standard comprised of 2 mg/ml each of 3-benzoylpyridine, Cortisone, 4-nitroaniline, and 4,4'-biphenol in methanol solution, designed for system performance applications.

  • Formulated as a liquid for UV detection.
  • Designed for SFC/UPC2 system types.
  • Quantitative reference standard.
  • Volume of 1 mL per package.
  • Shelf life of 36 months when stored at 4°C.
  • Shipped under ambient conditions.
  • Contains hazardous material requiring special freight handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.