Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Standards Kit, Qualification, System Performance, UV Detection

Catalog No. 502392995 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This kit provides a comprehensive set of standards for verifying the performance of UPC2 systems, including various caffeine, coumarin, flavone, thymine, prednisone solutions, and an erbium perchlorate solution.

  • Detection method: UV
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: SFC/UPC2
  • Instrument: SFx
  • Quantitative: Yes
  • Shelf life: 36 months
  • Shipping condition: Ambient
  • Volume: 10 mL per ampule
  • Type: Reference
  • Application: System performance
  • Units per package: 1 pk
  • Hazardous material: This product contains hazardous material which requires special freight handling; additional charges may apply.
  • Kit contents: Multiple 10 mL ampules of various standards for UPC2 system verification, including caffeine, coumarin, flavone, thymine, prednisone solutions, and erbium perchlorate.

Catalog No. 50-239-2995 Supplier Waters Corp Supplier No. 186006829
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit provides a comprehensive set of standards for verifying the performance of UPC2 systems, including various caffeine, coumarin, flavone, thymine, prednisone solutions, and an erbium perchlorate solution.

  • Detection method: UV
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: SFC/UPC2
  • Instrument: SFx
  • Quantitative: Yes
  • Shelf life: 36 months
  • Shipping condition: Ambient
  • Volume: 10 mL per ampule
  • Type: Reference
  • Application: System performance
  • Units per package: 1 pk
  • Hazardous material: This product contains hazardous material which requires special freight handling; additional charges may apply.
  • Kit contents: Multiple 10 mL ampules of various standards for UPC2 system verification, including caffeine, coumarin, flavone, thymine, prednisone solutions, and erbium perchlorate.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.