Learn More
Enzo Life Sciences UPF1035 (5mg). CAS: 370872-09-6
SDP
Supplier:  Enzo Life Sciences ALX270498M005
Encompass_Preferred
PARP-2 specific inhibitor with 60-fold selectivity for PARP-2 (IC50=0.15 ± 0.04μM) over PARP-1 (IC50=9.0 ± 0.7μM). Can be used for further characterization of PARP-2 in pathophysiological conditions. Alternative name: 5-Benzoyloxyisoquinolin-1(2H)-one. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8622