Learn More
Aviva Systems Biology Corp UPF2 Antibody - N-terminal region (ARP40814_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40814_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UPF2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human UPF2. Alternate names: DKFZp434D222, HUPF2, KIAA1408, MGC138834, MGC138835, RENT2, smg-3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-284-894