Learn More
Aviva Systems Biology Corp UPF3A Antibody - middle region (ARP41000_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41000_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UPF3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UPF3A. Alternate names: HUPF3A, RENT3A, UPF3ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57693