Learn More
TARGETMOL CHEMICALS INC UPGL00004 5MG
SDP
Catalog No. 502581114
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UPGL00004 is a potent glutaminase C (GAC) inhibitor with an IC50 of 29 nM. Kd of UPGL00004 for GAC was 27 nM. UPGL00004 has a strong inhibitory effect on the proliferation of highly aggressive triple negative breast cancer cell lines. purity: 97%
Catalog No. 50-258-1114 Supplier TARGETMOL CHEMICALS INC Supplier No. T172065MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UPGL00004 is a potent glutaminase C (GAC) inhibitor with an IC50 of 29 nM. Kd of UPGL00004 for GAC was 27 nM. UPGL00004 has a strong inhibitory effect on the proliferation of highly aggressive triple negative breast cancer cell lines. purity: 97%