Learn More
Anton Paar Upgradable monomode microwave reactor (850 W) for applications up to 260 °C and 20 bar. Compatible with borosilicate glass or SiC vials. Features touchscreen, magnetic stirring, pressure and IR sensors, plus USB and Ethernet connectivity.
Catalog No. 5000580484
Encompass_Preferred
Power supply: 230V, 50 Hz/60 Hz, 1600 VAIncluded:- Spill bin (83740)- 5 pcs. Reaction Vial G10- 2 pcs. Reaction vial G30- 7 pcs. Snap cap- 20 pcs. Silicon septum- 10 pcs. Stir bar PTFE 10x3- Blind pin (89189)- Adaptor ring (88999)- 2 pcs. Cavity collar (83212)- 3 pcs. Filter disc (84294)- Piston cover key (89196)
Catalog No. 50-005-80484 Supplier Anton Paar Supplier No. 163918
Please contact your sales representative for a quote or further ordering instructions.
Power supply: 230V, 50 Hz/60 Hz, 1600 VAIncluded:- Spill bin (83740)- 5 pcs. Reaction Vial G10- 2 pcs. Reaction vial G30- 7 pcs. Snap cap- 20 pcs. Silicon septum- 10 pcs. Stir bar PTFE 10x3- Blind pin (89189)- Adaptor ring (88999)- 2 pcs. Cavity collar (83212)- 3 pcs. Filter disc (84294)- Piston cover key (89196)