Learn More
Waters Corp Waters Corporation Upgrade Kit, Transducer BGM, for HPLC Systems
Catalog No. 50179759 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This upgrade kit is designed to update early 2545 Binary Gradient Modules in HPLC systems. It provides a newer style pressure transducer with a die-cast housing, along with tubing assemblies and an adapter, to replace older, round version transducers.
  • Provides one pressure transducer and tubing assemblies.
  • Updates early 2545 Binary Gradient Modules.
  • Features a newer style transducer with a die-cast housing.
  • Replaces older, round version transducers.
  • Includes an adapter for installation to the pump head.
  • Suitable for HPLC systems.
  • Units per package: 1.
Catalog No. 50-179-759 Supplier Waters Corp Supplier No. 700004864
This upgrade kit is designed to update early 2545 Binary Gradient Modules in HPLC systems. It provides a newer style pressure transducer with a die-cast housing, along with tubing assemblies and an adapter, to replace older, round version transducers.
  • Provides one pressure transducer and tubing assemblies.
  • Updates early 2545 Binary Gradient Modules.
  • Features a newer style transducer with a die-cast housing.
  • Replaces older, round version transducers.
  • Includes an adapter for installation to the pump head.
  • Suitable for HPLC systems.
  • Units per package: 1.