Learn More
MSC Guardair Retrofit Kit, Drum-Top Vacuum Head, 5/10 Gallon, Wet/Dry
Catalog No. 502837603
Encompass
This retrofit kit is designed to convert existing Guardair 5 and 10 gallon vacuums into PowerQUAD vacuums. It offers several advantages including low air usage, high vacuum flow, high vacuum head, low noise levels, and true wet/dry vacuum capability.
  • Easily turns your existing 5 and 10 gallon vacuum into a PowerQUAD vacuum.
  • Offers low air usage, high vacuum flow, high vacuum head, and low noise levels.
  • Provides true wet dry vacuum capabilities.
  • Includes PowerQUAD vacuum engine.
  • Includes PowerQUAD vacuum engine gasket.
  • Includes PowerQUAD nozzle set.
  • Includes wire cage.
  • Includes standard cartridge filter.
  • Includes 5/10 gallon wire cage coalescing filter.
  • Includes ball float.
  • Includes installation hardware.
Catalog No. 50-283-7603 Supplier MSC Supplier No. 64362395
This retrofit kit is designed to convert existing Guardair 5 and 10 gallon vacuums into PowerQUAD vacuums. It offers several advantages including low air usage, high vacuum flow, high vacuum head, low noise levels, and true wet/dry vacuum capability.
  • Easily turns your existing 5 and 10 gallon vacuum into a PowerQUAD vacuum.
  • Offers low air usage, high vacuum flow, high vacuum head, and low noise levels.
  • Provides true wet dry vacuum capabilities.
  • Includes PowerQUAD vacuum engine.
  • Includes PowerQUAD vacuum engine gasket.
  • Includes PowerQUAD nozzle set.
  • Includes wire cage.
  • Includes standard cartridge filter.
  • Includes 5/10 gallon wire cage coalescing filter.
  • Includes ball float.
  • Includes installation hardware.