Learn More
MSC O-Cedar Scrub Brush, Polypropylene, 6 in Length, 12 Pack
Catalog No. 502846106
Encompass
This general-purpose scrubbing brush features a heavy-duty molded foam block designed for comfortable hand-held use. It is equipped with crimped polypropylene fibers, ensuring extra strength and enhanced fluid retention for effective cleaning across various applications, including upholstery.
  • Molded 6 in foam block fits comfortably in hand
  • Polypropylene bristles, 1 3/8 in, are crimped for strength and fluid retention
  • Overall length is 6 in with a 3 in brush width
  • Plastic handle and beige bristles
Catalog No. 50-284-6106 Supplier MSC Supplier No. 30590830
This general-purpose scrubbing brush features a heavy-duty molded foam block designed for comfortable hand-held use. It is equipped with crimped polypropylene fibers, ensuring extra strength and enhanced fluid retention for effective cleaning across various applications, including upholstery.
  • Molded 6 in foam block fits comfortably in hand
  • Polypropylene bristles, 1 3/8 in, are crimped for strength and fluid retention
  • Overall length is 6 in with a 3 in brush width
  • Plastic handle and beige bristles