Learn More
Ibidi USA -Plate 24 Well Black ID 14 mm
SDP
Catalog No. NC2124784
Encompass
Complete solution for wound healing experiments, requiring only a few steps from sample preparation to image analysisReproducible experiments owing to: a defined 500 µm cell free gap, no leakage during cultivation, and no material being left behind after the insert’s removal. For screening applications. High parallelization. Based on µ-Plate 24 Well Black ID 14 mm – i.e., suitable for fluorescence and high resolution microscopy. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. NC2124784 Supplier Ibidi USA Supplier No. 82426
Complete solution for wound healing experiments, requiring only a few steps from sample preparation to image analysisReproducible experiments owing to: a defined 500 µm cell free gap, no leakage during cultivation, and no material being left behind after the insert’s removal. For screening applications. High parallelization. Based on µ-Plate 24 Well Black ID 14 mm – i.e., suitable for fluorescence and high resolution microscopy. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box