Learn More
Waters Corp Waters Application Kit, UPLC AAA, UV Detection, 250 Samples
Catalog No. 50465949 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This UPLC AAA H-Class Applications Kit is designed for Amino Acid Analysis (AAA), providing UV detection and enough analysis for 250 samples. It is available in liquid and solid forms and should be refrigerated between 2°C and 8°C. The kit is based on a leading chemistry solution for accurate, high-sensitivity UPLC analysis, utilizing a reagent for single-step derivatization of primary and secondary amino acids.
  • Designed for Amino Acid Analysis (AAA).
  • Includes a derivatization kit.
  • Offers UV detection for 250 samples.
  • Available in liquid and solid forms.
  • Requires refrigeration between 2°C and 8°C.
  • Optimized for accurate, high-sensitivity UPLC analysis.
  • Utilizes a reagent for single-step amino acid derivatization.
  • Contains tested reagents, columns, and eluents.
Catalog No. 50-465-949 Supplier Waters Corp Supplier No. 176002983
Add to Cart
Add to Cart
Request a Quote
This UPLC AAA H-Class Applications Kit is designed for Amino Acid Analysis (AAA), providing UV detection and enough analysis for 250 samples. It is available in liquid and solid forms and should be refrigerated between 2°C and 8°C. The kit is based on a leading chemistry solution for accurate, high-sensitivity UPLC analysis, utilizing a reagent for single-step derivatization of primary and secondary amino acids.
  • Designed for Amino Acid Analysis (AAA).
  • Includes a derivatization kit.
  • Offers UV detection for 250 samples.
  • Available in liquid and solid forms.
  • Requires refrigeration between 2°C and 8°C.
  • Optimized for accurate, high-sensitivity UPLC analysis.
  • Utilizes a reagent for single-step amino acid derivatization.
  • Contains tested reagents, columns, and eluents.