Learn More
Waters Corp Waters CORTECS C18+ Method Validation Kit, 90Å, 1.6 μm, 2.1 mm x 100 mm
Catalog No. NC1679050 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three high-efficiency C18+ columns. Based on solid-core particles, these reversed-phase columns are designed to deliver excellent peak shape for basic analytes at low pH. The positively charged surface and C18 ligand offer higher retention and stability.
  • Solid-core particles improve performance by consistent packing and heat transfer, reducing A, B, and C terms.
  • Increased throughput through shorter column lengths, faster re-equilibration, and higher flow rates.
  • High resolution selectivity allows for quicker method construction.
  • Robustness: withstands temperatures up to 45°C, pressures up to 18,000 psi (1240 bar), and pH range of 2 to 8.
Catalog No. NC1679050 Supplier Waters Corp Supplier No. 186007179
This method validation kit includes three high-efficiency C18+ columns. Based on solid-core particles, these reversed-phase columns are designed to deliver excellent peak shape for basic analytes at low pH. The positively charged surface and C18 ligand offer higher retention and stability.
  • Solid-core particles improve performance by consistent packing and heat transfer, reducing A, B, and C terms.
  • Increased throughput through shorter column lengths, faster re-equilibration, and higher flow rates.
  • High resolution selectivity allows for quicker method construction.
  • Robustness: withstands temperatures up to 45°C, pressures up to 18,000 psi (1240 bar), and pH range of 2 to 8.