Learn More
Waters Corp Waters Corporation UPLC Melamine Analysis Package, Food Safety Screening, Low ppb Detection
Catalog No. 50780059 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The UPLC Melamine Analysis Package is an application kit for screening melamine and related compounds in food matrices like infant formulas and dairy products. It includes Oasis cartridges, vials, filters, and a choice of LC columns (HPLC or UPLC). Optimized for rapid, precise analytical results, it detects melamine at parts per billion (ppb) levels, supporting rigorous food safety testing.
  • Screens melamine and related compounds in food products.
  • Supports analysis in infant formulas and dairy products.
  • Uses Oasis cartridges as specified in US FDA LIB No. 4422.
  • Includes vials and filters for comprehensive testing.
  • Offers choice of HPLC or UPLC columns.
  • Provides rapid and accurate detection down to low ppb levels.
Catalog No. 50-780-059 Supplier Waters Corp Supplier No. 176001791
The UPLC Melamine Analysis Package is an application kit for screening melamine and related compounds in food matrices like infant formulas and dairy products. It includes Oasis cartridges, vials, filters, and a choice of LC columns (HPLC or UPLC). Optimized for rapid, precise analytical results, it detects melamine at parts per billion (ppb) levels, supporting rigorous food safety testing.
  • Screens melamine and related compounds in food products.
  • Supports analysis in infant formulas and dairy products.
  • Uses Oasis cartridges as specified in US FDA LIB No. 4422.
  • Includes vials and filters for comprehensive testing.
  • Offers choice of HPLC or UPLC columns.
  • Provides rapid and accurate detection down to low ppb levels.