Learn More
Waters Corp Waters Corporation UPLC Method Development Scouting Kit, 3.0 mm x 50 mm, 4-pack
Catalog No. 50791060 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This UPLC Method Development Scouting Kit facilitates efficient chromatographic method development, offering columns that reliably deliver desired separation results with broad selectivity.
  • Widest selectivity for method development at low and high pH.
  • Ideal for low ionic strength additives like formic acid.
  • Maximizes separation selectivity across diverse mobile-phase pH.
  • Accommodates retention of polar compounds.
  • Features C18 columns with varying silanol activity and hydrophobicity.
  • Combines RP and HILIC stationary phases for broad polarity analyte retention.
  • Available in multiple kit configurations, including 4-pack or 2-pack options.
Catalog No. 50-791-060 Supplier Waters Corp Supplier No. 176001883
Add to Cart
Add to Cart
Request a Quote
This UPLC Method Development Scouting Kit facilitates efficient chromatographic method development, offering columns that reliably deliver desired separation results with broad selectivity.
  • Widest selectivity for method development at low and high pH.
  • Ideal for low ionic strength additives like formic acid.
  • Maximizes separation selectivity across diverse mobile-phase pH.
  • Accommodates retention of polar compounds.
  • Features C18 columns with varying silanol activity and hydrophobicity.
  • Combines RP and HILIC stationary phases for broad polarity analyte retention.
  • Available in multiple kit configurations, including 4-pack or 2-pack options.