Learn More
Waters Corp WATERS CORPORATION UPLC Method Development Scouting Kit, 2.1 mm, 50 mm, 4/pk
Catalog No. 50791017 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The UPLC method development scouting kit is designed to accommodate method development approaches by combining 1.7 μm BEH C18, Shield RP18, Phenyl, and 1.8 μm HSS T3 column chemistries. This enables efficient and effective method development. Utilizing UPLC Technology for method development can significantly improve throughput, reduce cost per sample, and decrease analysis time while maintaining or improving separation integrity. This ultimately helps bring products to market faster and improves the overall profitability of the assay.
  • Combines 1.7 μm BEH C18, Shield RP18, Phenyl, and 1.8 μm HSS T3 column chemistries.
  • Aids in efficient and effective method development.
  • Utilizes UPLC Technology for improved throughput.
  • Reduces cost per sample and analysis time.
  • Maintains or improves separation integrity.
Catalog No. 50-791-017 Supplier Waters Corp Supplier No. 176001603
Add to Cart
Add to Cart
Request a Quote
The UPLC method development scouting kit is designed to accommodate method development approaches by combining 1.7 μm BEH C18, Shield RP18, Phenyl, and 1.8 μm HSS T3 column chemistries. This enables efficient and effective method development. Utilizing UPLC Technology for method development can significantly improve throughput, reduce cost per sample, and decrease analysis time while maintaining or improving separation integrity. This ultimately helps bring products to market faster and improves the overall profitability of the assay.
  • Combines 1.7 μm BEH C18, Shield RP18, Phenyl, and 1.8 μm HSS T3 column chemistries.
  • Aids in efficient and effective method development.
  • Utilizes UPLC Technology for improved throughput.
  • Reduces cost per sample and analysis time.
  • Maintains or improves separation integrity.