Learn More
Waters Corp VanGuard Pre-column, C18, 100Å, 1.8 μm, 2.1 mm
Catalog No. NC3254856 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC Peptide HSS T3 VanGuard Pre-column is designed to protect and extend the lifetime of analytical columns without compromising chromatographic performance. It uses HSS T3 particles, suitable for separations requiring silica-based selectivities or increased retention of hydrophilic peptides. This pre-column helps filter out chemical or particle contaminants from the mobile phase stream, thereby increasing the analytical performance of the column.
  • Protects and prolongs analytical column lifetimes.
  • Maintains chromatographic performance.
  • HSS T3 particles provide silica-based selectivities and increased retention of hydrophilic peptides.
  • Filters chemical or particle contaminants from the mobile phase.
  • Manufactured to cGMP and ISO 9001 guidelines.
  • Extends UPLC analytical column life across UPLC, UHPLC, and HPLC systems.
  • Tested and approved for intended usage.
Catalog No. NC3254856 Supplier Waters Corp Supplier No. 186008757
The ACQUITY UPLC Peptide HSS T3 VanGuard Pre-column is designed to protect and extend the lifetime of analytical columns without compromising chromatographic performance. It uses HSS T3 particles, suitable for separations requiring silica-based selectivities or increased retention of hydrophilic peptides. This pre-column helps filter out chemical or particle contaminants from the mobile phase stream, thereby increasing the analytical performance of the column.
  • Protects and prolongs analytical column lifetimes.
  • Maintains chromatographic performance.
  • HSS T3 particles provide silica-based selectivities and increased retention of hydrophilic peptides.
  • Filters chemical or particle contaminants from the mobile phase.
  • Manufactured to cGMP and ISO 9001 guidelines.
  • Extends UPLC analytical column life across UPLC, UHPLC, and HPLC systems.
  • Tested and approved for intended usage.