Learn More
Waters Corp Waters Method Development Kit, UPLC, Therapeutic Peptide
Catalog No. 50465905 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The UPLC Therapeutic Peptide Method Development kit simplifies the process of sample preparation and LC method development for the analysis of therapeutic peptides in plasma.
  • One 96-well Oasis Peptide μElution Method Development Plate
  • One ACQUITY UPLC 1.7 μm BEH 300 C18 reversed-phase 2.1x50 mm column
  • Three 96-well 1 mL collection plates and cap mats
  • A detailed screening protocol
Catalog No. 50-465-905 Supplier Waters Corp Supplier No. 176001835
The UPLC Therapeutic Peptide Method Development kit simplifies the process of sample preparation and LC method development for the analysis of therapeutic peptides in plasma.
  • One 96-well Oasis Peptide μElution Method Development Plate
  • One ACQUITY UPLC 1.7 μm BEH 300 C18 reversed-phase 2.1x50 mm column
  • Three 96-well 1 mL collection plates and cap mats
  • A detailed screening protocol