Learn More
Meso Scale Discovery UPLEX Adipokine Combo 1 ms
SDP
Catalog No. 5000082205
Encompass
The Meso Scale Discovery U-PLEX Adipokine Combo 1 Mouse Assay is a multiplex immunoassay kit that allows simultaneous measurement of mouse adiponectin leptin PAI-1 total and resistin It uses electrochemiluminescence detection technology on Meso Scale Discovery s QuickPlex SQ 120 instrument The assay requires 50 L of sample per well and can be completed in 3 5 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer This assay provides a sensitive and reproducible method for analyzing adipokines in mouse serum plasma and cell culture supernatants
Catalog No. 50-000-82205 Supplier Meso Scale Discovery Supplier No. K15299K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Adipokine Combo 1 Mouse Assay is a multiplex immunoassay kit that allows simultaneous measurement of mouse adiponectin leptin PAI-1 total and resistin It uses electrochemiluminescence detection technology on Meso Scale Discovery s QuickPlex SQ 120 instrument The assay requires 50 L of sample per well and can be completed in 3 5 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer This assay provides a sensitive and reproducible method for analyzing adipokines in mouse serum plasma and cell culture supernatants