Learn More
Meso Scale Discovery U-PLEX Adipokine Combo 1 (hu) SECTOR (25 PL)
SDP
Catalog No. NC3786783
Encompass
U-PLEX Combos offer popular panels of biomarkers that have been grouped together for convenient ordering. They have been tested for multiplex compatibility and are fulfilled as a kit. The U-PLEX Adipokine Combo 1 (human) combines ten assays (Table 1) that are a mix of traditional metabolic markers (Insulin and Leptin), emerging biomarkers in metabolic syndrome (BDNF and β-NGF), and several biomarkers related to the inflammatory status of tissue. The assays can be used for quantitative measurements in the sample types listed in the "Specifications" tab below, and offer detection limits (LLODs) in the sub- to low pg/mL range.
Catalog No. NC3786783 Supplier Meso Scale Discovery Supplier No. K15276K4
Add to Cart
Add to Cart
Request a Quote
U-PLEX Combos offer popular panels of biomarkers that have been grouped together for convenient ordering. They have been tested for multiplex compatibility and are fulfilled as a kit. The U-PLEX Adipokine Combo 1 (human) combines ten assays (Table 1) that are a mix of traditional metabolic markers (Insulin and Leptin), emerging biomarkers in metabolic syndrome (BDNF and β-NGF), and several biomarkers related to the inflammatory status of tissue. The assays can be used for quantitative measurements in the sample types listed in the "Specifications" tab below, and offer detection limits (LLODs) in the sub- to low pg/mL range.