Learn More
Meso Scale Discovery UPLEX ALZ DIS Combo 1HMN
SDP
Catalog No. 5000079280
Encompass
The Meso Scale Discovery U-PLEX Alzheimer s Disease Combo 1 Human Kit is a multiplex immunoassay measuring amyloid beta 40 amyloid beta 42 total tau and phosphorylated tau 181 in human serum plasma and CSF Using MSD s QuickPlex SQ 120 instrument it offers a sensitive method for assessing Alzheimer s biomarkers With a streamlined workflow it enables efficient profiling and monitoring of disease progression and therapeutic response delivering robust data from just 50 L of sample
Catalog No. 50-000-79280 Supplier Meso Scale Discovery Supplier No. K15683K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Alzheimer s Disease Combo 1 Human Kit is a multiplex immunoassay measuring amyloid beta 40 amyloid beta 42 total tau and phosphorylated tau 181 in human serum plasma and CSF Using MSD s QuickPlex SQ 120 instrument it offers a sensitive method for assessing Alzheimer s biomarkers With a streamlined workflow it enables efficient profiling and monitoring of disease progression and therapeutic response delivering robust data from just 50 L of sample