Learn More
Meso Scale Discovery UPLEX ALZ DIS Combo 1HMN
SDP
Catalog No. 5000080863
Encompass
The Meso Scale Discovery U-PLEX Alzheimer s Disease Combo 1 Human Kit is a multiplex immunoassay kit that allows simultaneous measurement of amyloid beta 40 amyloid beta 42 total tau and phosphorylated tau 181 in human samples such as CSF serum and plasma This kit uses electrochemiluminescent detection technology and provides reagents for 96 tests when using the standard assay protocol The kit can be used to aid in the differential diagnosis of Alzheimer s disease by measuring changes in levels of the 4 key biomarkers It is for research use only
Catalog No. 50-000-80863 Supplier Meso Scale Discovery Supplier No. K15683K1
The Meso Scale Discovery U-PLEX Alzheimer s Disease Combo 1 Human Kit is a multiplex immunoassay kit that allows simultaneous measurement of amyloid beta 40 amyloid beta 42 total tau and phosphorylated tau 181 in human samples such as CSF serum and plasma This kit uses electrochemiluminescent detection technology and provides reagents for 96 tests when using the standard assay protocol The kit can be used to aid in the differential diagnosis of Alzheimer s disease by measuring changes in levels of the 4 key biomarkers It is for research use only