Learn More
Meso Scale Discovery UPLEX ALZ DIS Combo 1HMN
SDP
Catalog No. 5000081056
Encompass
The Meso Scale Discovery U-PLEX Alzheimer s Disease Combo 1 Human Kit is a multiplex immunoassay measuring amyloid beta 40 amyloid beta 42 total tau and phosphorylated tau 181 in human serum plasma and CSF It uses MSD s electrochemiluminescent detection for sensitive biomarker measurement The kit includes a 96-well plate antibodies calibrators controls diluent and buffers for one plate Recommended sample volume is 50 L Testing takes 3 5 hours with a total time of 4 5 hours aiding Alzheimer s research and drug development
Catalog No. 50-000-81056 Supplier Meso Scale Discovery Supplier No. K15683K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Alzheimer s Disease Combo 1 Human Kit is a multiplex immunoassay measuring amyloid beta 40 amyloid beta 42 total tau and phosphorylated tau 181 in human serum plasma and CSF It uses MSD s electrochemiluminescent detection for sensitive biomarker measurement The kit includes a 96-well plate antibodies calibrators controls diluent and buffers for one plate Recommended sample volume is 50 L Testing takes 3 5 hours with a total time of 4 5 hours aiding Alzheimer s research and drug development