Learn More
Meso Scale Discovery UPLEX Angiogenesis Combo hu
SDP
Catalog No. 5000078314
Encompass
The Meso Scale Discovery U-PLEX Angiogenesis Combo 1 Human Kit is a multiplex immunoassay that measures VEGF-A VEGF-C VEGF-D PlGF and sFlt-1 in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer This product enables the quantification of multiple angiogenesis-related analytes from a single small-volume sample
Catalog No. 50-000-78314 Supplier Meso Scale Discovery Supplier No. K15339K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Angiogenesis Combo 1 Human Kit is a multiplex immunoassay that measures VEGF-A VEGF-C VEGF-D PlGF and sFlt-1 in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer This product enables the quantification of multiple angiogenesis-related analytes from a single small-volume sample