Learn More
Meso Scale Discovery UPLEX CART Cell Combo 2HMN
SDP
Catalog No. 5000082452
Encompass
The U-PLEX CAR-T Cell Combo 2 Human Sector 1 Panel Kit from Meso Scale Discovery is a multiplex immunoassay kit for detecting human cytokines and chemokines It includes a 96-well plate with capture and detection antibodies Read Buffer and calibrator solutions It measures IL-2 IL-4 IL-6 IL-8 IL-10 IL-13 IFN- TNF- MIP-1 MIP-1 and RANTES from various samples The electrochemiluminescent method offers high sensitivity and a wide dynamic range ideal for CAR-T cell therapy and immunotherapy research
Catalog No. 50-000-82452 Supplier Meso Scale Discovery Supplier No. K15600K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX CAR-T Cell Combo 2 Human Sector 1 Panel Kit from Meso Scale Discovery is a multiplex immunoassay kit for detecting human cytokines and chemokines It includes a 96-well plate with capture and detection antibodies Read Buffer and calibrator solutions It measures IL-2 IL-4 IL-6 IL-8 IL-10 IL-13 IFN- TNF- MIP-1 MIP-1 and RANTES from various samples The electrochemiluminescent method offers high sensitivity and a wide dynamic range ideal for CAR-T cell therapy and immunotherapy research