Learn More
Meso Scale Discovery UPLEX Chemokine Combo 1 ms
SDP
Catalog No. 5000082135
Encompass
The U-PLEX Chemokine Combo 1 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous measurement of 10 mouse chemokines (CCL2 CCL3 CCL4 CCL5 CCL11 CXCL1 CXCL2 CXCL5 CXCL9 CXCL10) from cell culture supernatants serum plasma (EDTA heparin) and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required for the assay
Catalog No. 50-000-82135 Supplier Meso Scale Discovery Supplier No. K15321K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Chemokine Combo 1 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous measurement of 10 mouse chemokines (CCL2 CCL3 CCL4 CCL5 CCL11 CXCL1 CXCL2 CXCL5 CXCL9 CXCL10) from cell culture supernatants serum plasma (EDTA heparin) and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required for the assay