Learn More
Meso Scale Discovery UPLEX Chemokine Combo 1 NHP
SDP
Catalog No. 5000078252
Encompass
The U-PLEX Chemokine Combo 1 NHP is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous measurement of 10 non-human primate chemokines (CCL2 CCL3 CCL4 CCL5 CXCL8 CXCL9 CXCL10 CXCL11 CXCL12 and CX3CL1) in serum plasma and culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology provides high sensitivity and a broad dynamic range requires low sample volume (25-50 L) and has a short 2 5 hour total assay time The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls
Catalog No. 50-000-78252 Supplier Meso Scale Discovery Supplier No. K15055K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Chemokine Combo 1 NHP is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous measurement of 10 non-human primate chemokines (CCL2 CCL3 CCL4 CCL5 CXCL8 CXCL9 CXCL10 CXCL11 CXCL12 and CX3CL1) in serum plasma and culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology provides high sensitivity and a broad dynamic range requires low sample volume (25-50 L) and has a short 2 5 hour total assay time The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls