Learn More
Meso Scale Discovery UPLEX Chemokine Combo 1 NHP
SDP
Catalog No. 5000081594
Encompass
The U-PLEX Chemokine Combo 1 NHP Assay is a multiplex immunoassay panel that simultaneously measures the concentrations of 10 chemokine analytes in non-human primate samples CCL2 CCL3 CCL4 CCL5 CXCL8 CXCL9 CXCL10 CXCL11 CXCL12 and CX3CL1 This assay uses electrochemiluminescence detection technology on the Meso Scale Discovery platform It requires 50 L of sample per well and can be used with serum plasma and other biological fluids The panel allows efficient high-throughput quantification of multiple chemokine biomarkers related to inflammation immune response and chemotaxis It provides reproducible sensitive results across a broad dynamic range
Catalog No. 50-000-81594 Supplier Meso Scale Discovery Supplier No. K15055K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Chemokine Combo 1 NHP Assay is a multiplex immunoassay panel that simultaneously measures the concentrations of 10 chemokine analytes in non-human primate samples CCL2 CCL3 CCL4 CCL5 CXCL8 CXCL9 CXCL10 CXCL11 CXCL12 and CX3CL1 This assay uses electrochemiluminescence detection technology on the Meso Scale Discovery platform It requires 50 L of sample per well and can be used with serum plasma and other biological fluids The panel allows efficient high-throughput quantification of multiple chemokine biomarkers related to inflammation immune response and chemotaxis It provides reproducible sensitive results across a broad dynamic range