Learn More
Meso Scale Discovery UPLEX Chemokine Combo ms
SDP
Catalog No. 5000078319
Encompass
The U-PLEX Chemokine Combo Mouse Assay is a multiplex immunoassay kit that allows simultaneous measurement of 10 mouse chemokines (CCL2 CCL3 CCL4 CCL5 CCL11 CXCL1 CXCL2 CXCL5 CXCL9 CXCL10) from cell culture supernatants serum plasma (EDTA heparin) and other biological fluids in a single sample volume of 50 L The assay utilizes Meso Scale Discovery electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with minimal sample consumption The kit includes a 10-plex mouse standard and all reagents required to perform the assay
Catalog No. 50-000-78319 Supplier Meso Scale Discovery Supplier No. K15099K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Chemokine Combo Mouse Assay is a multiplex immunoassay kit that allows simultaneous measurement of 10 mouse chemokines (CCL2 CCL3 CCL4 CCL5 CCL11 CXCL1 CXCL2 CXCL5 CXCL9 CXCL10) from cell culture supernatants serum plasma (EDTA heparin) and other biological fluids in a single sample volume of 50 L The assay utilizes Meso Scale Discovery electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with minimal sample consumption The kit includes a 10-plex mouse standard and all reagents required to perform the assay