Learn More
Meso Scale Discovery UPLEX Cytokine Combo 1 NHP
SDP
Catalog No. 5000079194
Encompass
The U-PLEX Cytokine Combo 1 NHP Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of 15 non-human primate cytokines chemokines and growth factors in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and stop solution The assay uses electrochemiluminescence for high sensitivity and broad dynamic range This kit allows efficient and accurate analysis of NHP samples for immunology and inflammation research
Catalog No. 50-000-79194 Supplier Meso Scale Discovery Supplier No. K15057K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Cytokine Combo 1 NHP Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of 15 non-human primate cytokines chemokines and growth factors in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and stop solution The assay uses electrochemiluminescence for high sensitivity and broad dynamic range This kit allows efficient and accurate analysis of NHP samples for immunology and inflammation research