Learn More
Meso Scale Discovery UPLEX Development Pack2ASY ate
SDP
Catalog No. 5000082167
Encompass
The Meso Scale Discovery U-PLEX Development Pack 2 Assay is a multiplex immunoassay kit that allows users to develop custom assays for up to 2 biomarkers It contains the components needed to develop and optimize an immunoassay including plates linkers detection antibodies buffers calibrators and controls The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample consumption This product enables the quantification of protein biomarkers for life science research applications
Catalog No. 50-000-82167 Supplier Meso Scale Discovery Supplier No. K15227N1
The Meso Scale Discovery U-PLEX Development Pack 2 Assay is a multiplex immunoassay kit that allows users to develop custom assays for up to 2 biomarkers It contains the components needed to develop and optimize an immunoassay including plates linkers detection antibodies buffers calibrators and controls The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample consumption This product enables the quantification of protein biomarkers for life science research applications