Learn More
Meso Scale Discovery UPLEX Development Pack3842ASY
SDP
Catalog No. 5000079420
Encompass
The Meso Scale Discovery U-PLEX Development Pack 384-Well 2-Assay is a multiplex immunoassay kit for developing custom assays to measure up to 2 biomarkers in 384-well plates It includes plates linkers detection antibodies buffers calibrators controls and accessories for up to 2 custom U-PLEX assays Users can choose capture and detection antibodies The electrochemiluminescent detection offers high sensitivity a dynamic range of up to 5 logs and low background aiding in the rapid development and optimization of biomarker assays
Catalog No. 50-000-79420 Supplier Meso Scale Discovery Supplier No. K25227N4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Development Pack 384-Well 2-Assay is a multiplex immunoassay kit for developing custom assays to measure up to 2 biomarkers in 384-well plates It includes plates linkers detection antibodies buffers calibrators controls and accessories for up to 2 custom U-PLEX assays Users can choose capture and detection antibodies The electrochemiluminescent detection offers high sensitivity a dynamic range of up to 5 logs and low background aiding in the rapid development and optimization of biomarker assays