Learn More
Meso Scale Discovery UPLEX Development Pack3842ASY
SDP
Catalog No. 5000080694
Encompass
The Meso Scale Discovery U-PLEX Development Pack 384-Well 2-Assay is a multiplex immunoassay kit for developing custom assays on the MSD platform It contains a 384-well MULTI-ARRAY plate 2 vials of Read Buffer T (4X) and reagents for developing and optimizing up to 2 custom assays The electrochemiluminescent detection allows for high sensitivity and a large dynamic range This kit enables assay development for biomarkers immunogenicity and pharmacodynamics on MSD s high-throughput 384-well platform With optimized assays the kit can enable quantification of proteins peptides antibodies and other analytes in biological samples
Catalog No. 50-000-80694 Supplier Meso Scale Discovery Supplier No. K25227N2
The Meso Scale Discovery U-PLEX Development Pack 384-Well 2-Assay is a multiplex immunoassay kit for developing custom assays on the MSD platform It contains a 384-well MULTI-ARRAY plate 2 vials of Read Buffer T (4X) and reagents for developing and optimizing up to 2 custom assays The electrochemiluminescent detection allows for high sensitivity and a large dynamic range This kit enables assay development for biomarkers immunogenicity and pharmacodynamics on MSD s high-throughput 384-well platform With optimized assays the kit can enable quantification of proteins peptides antibodies and other analytes in biological samples