Learn More
Meso Scale Discovery UPLEX Development Pack3843ASY
SDP
Catalog No. 5000079421
Encompass
The Meso Scale Discovery U-PLEX Development Pack 384-Well 3-Assay (K25228N) is a multiplex immunoassay kit that allows users to develop up to 3 custom assays in a 384-well plate format The kit contains a 384-well plate detection antibodies linkers buffers calibrators controls and substrates sufficient for developing and testing 3 custom assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low background This kit is intended for research use only to develop custom assays for biomarkers of interest
Catalog No. 50-000-79421 Supplier Meso Scale Discovery Supplier No. K25228N2
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Development Pack 384-Well 3-Assay (K25228N) is a multiplex immunoassay kit that allows users to develop up to 3 custom assays in a 384-well plate format The kit contains a 384-well plate detection antibodies linkers buffers calibrators controls and substrates sufficient for developing and testing 3 custom assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low background This kit is intended for research use only to develop custom assays for biomarkers of interest