Learn More
Meso Scale Discovery UPLEX Development Pack3843ASY
SDP
Catalog No. 5000080443
Encompass
The Meso Scale Discovery U-PLEX Development Pack 384-Well 3 Assay Kit is a multiplex immunoassay kit that allows users to develop up to 3 custom assays in a 384-well plate format The kit contains a 384-well plate pre-coated with patterned carbon electrodes a vial of Read Buffer vials of Detection Antibody Diluent vials of 10X Wash Buffer a plate seal and SULFO-TAG NHS-Ester labels for antibody conjugation The electrochemiluminescent detection allows for high sensitivity and a large dynamic range This kit enables efficient custom assay development for biomarker research
Catalog No. 50-000-80443 Supplier Meso Scale Discovery Supplier No. K25228N4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Development Pack 384-Well 3 Assay Kit is a multiplex immunoassay kit that allows users to develop up to 3 custom assays in a 384-well plate format The kit contains a 384-well plate pre-coated with patterned carbon electrodes a vial of Read Buffer vials of Detection Antibody Diluent vials of 10X Wash Buffer a plate seal and SULFO-TAG NHS-Ester labels for antibody conjugation The electrochemiluminescent detection allows for high sensitivity and a large dynamic range This kit enables efficient custom assay development for biomarker research