Learn More
Meso Scale Discovery UPLEX Development Pack3844ASY
SDP
Catalog No. 5000079434
Encompass
The Meso Scale Discovery U-PLEX Development Pack 384-Well 4-Assay is a multiplex immunoassay kit for developing custom assays on the MSD platform It contains 4 plates with 384 carbon electrodes per well allowing up to 4 custom assays per plate It also includes detection antibodies linkers buffers calibrators controls and plates seals needed to develop and optimize the assays The carbon electrode plates are compatible with MSD s imaging systems for electrochemiluminescent detection This kit enables efficient custom assay development for biomarkers cytokines and other analytes
Catalog No. 50-000-79434 Supplier Meso Scale Discovery Supplier No. K25229N4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Development Pack 384-Well 4-Assay is a multiplex immunoassay kit for developing custom assays on the MSD platform It contains 4 plates with 384 carbon electrodes per well allowing up to 4 custom assays per plate It also includes detection antibodies linkers buffers calibrators controls and plates seals needed to develop and optimize the assays The carbon electrode plates are compatible with MSD s imaging systems for electrochemiluminescent detection This kit enables efficient custom assay development for biomarkers cytokines and other analytes