Learn More
Meso Scale Discovery UPLEX Development Pack3ASY ate
SDP
Catalog No. 5000081324
Encompass
The Meso Scale Discovery U-PLEX Development Pack 3 Assay is a multiplex immunoassay kit for developing custom assays to measure up to 3 biomarkers from a single sample It includes a 10-spot MULTI-ARRAY 96-well plate SULFO-TAG NHS-Ester labels for antibody conjugation assay diluent read buffer wash buffer blocker solutions and plate seals Ideal for research in cancer immunology and neurobiology it offers high sensitivity and rapid results using 50-100 L of sample per well with electrochemiluminescence detection
Catalog No. 50-000-81324 Supplier Meso Scale Discovery Supplier No. K15228N4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Development Pack 3 Assay is a multiplex immunoassay kit for developing custom assays to measure up to 3 biomarkers from a single sample It includes a 10-spot MULTI-ARRAY 96-well plate SULFO-TAG NHS-Ester labels for antibody conjugation assay diluent read buffer wash buffer blocker solutions and plate seals Ideal for research in cancer immunology and neurobiology it offers high sensitivity and rapid results using 50-100 L of sample per well with electrochemiluminescence detection