Learn More
Meso Scale Discovery UPLEX Development Pack4ASY ate
SDP
Catalog No. 5000082168
Encompass
The Meso Scale Discovery U-PLEX Development Pack 4 Assay is a multiplex immunoassay kit that allows users to develop up to 4 custom assays for biomarkers of interest The kit contains plates linkers detection antibodies buffers calibrators and controls needed to develop and run the assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements Assays developed with this kit are for research use only
Catalog No. 50-000-82168 Supplier Meso Scale Discovery Supplier No. K15229N1
The Meso Scale Discovery U-PLEX Development Pack 4 Assay is a multiplex immunoassay kit that allows users to develop up to 4 custom assays for biomarkers of interest The kit contains plates linkers detection antibodies buffers calibrators and controls needed to develop and run the assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements Assays developed with this kit are for research use only