Learn More
Meso Scale Discovery UPLEX Development Pack5ASY ate
SDP
Catalog No. 5000079990
Encompass
The Meso Scale Discovery U-PLEX Development Pack 5 Assay is a customizable multiplex immunoassay kit that allows users to develop up to 5 biomarker assays of their choice on Meso Scale Discovery s unique electrochemiluminescent detection platform The kit contains all necessary reagents and plates to develop and optimize custom assays including linkers blockers read buffer wash buffer diluent and standard cytokines The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements Once optimized assays developed with this kit allow rapid reproducible analysis of biomarkers from a variety of sample types
Catalog No. 50-000-79990 Supplier Meso Scale Discovery Supplier No. K15230N1
The Meso Scale Discovery U-PLEX Development Pack 5 Assay is a customizable multiplex immunoassay kit that allows users to develop up to 5 biomarker assays of their choice on Meso Scale Discovery s unique electrochemiluminescent detection platform The kit contains all necessary reagents and plates to develop and optimize custom assays including linkers blockers read buffer wash buffer diluent and standard cytokines The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements Once optimized assays developed with this kit allow rapid reproducible analysis of biomarkers from a variety of sample types